Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614349 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF638 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF638 - zinc finger protein 638 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014972.2 | Intron | NP_001014972.1 | ||||
NM_001252612.1 | Intron | NP_001239541.1 | ||||
NM_001252613.1 | Intron | NP_001239542.1 | ||||
NM_014497.4 | Intron | NP_055312.2 | ||||
XM_005264263.1 | Intron | XP_005264320.1 | ||||
XM_006711989.1 | Intron | XP_006712052.1 | ||||
XM_011532768.2 | Intron | XP_011531070.2 | ||||
XM_017003809.1 | Intron | XP_016859298.1 | ||||
XM_017003810.1 | Intron | XP_016859299.1 | ||||
XM_017003811.1 | Intron | XP_016859300.1 | ||||
XM_017003812.1 | Intron | XP_016859301.1 |