Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611356 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GKAP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GKAP1 - G kinase anchoring protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135953.1 | Intron | NP_001129425.1 | ||||
NM_025211.3 | Intron | NP_079487.2 | ||||
XM_005252241.2 | Intron | XP_005252298.1 | ||||
XM_006717293.1 | Intron | XP_006717356.1 | ||||
XM_011519058.2 | Intron | XP_011517360.1 | ||||
XM_017015171.1 | Intron | XP_016870660.1 |
Set Membership: |
HapMap |