Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGTGGGCCATCAGCAGCCAGAGG[A/T]GCTCCTGAATCTAAACAGTATCCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610855 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD26 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD26 - ankyrin repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256053.1 | 23931 | Intron | NP_001242982.1 | |||
NM_014915.2 | 23931 | Intron | NP_055730.2 | |||
XM_006717423.2 | 23931 | Intron | XP_006717486.1 | |||
XM_006717425.3 | 23931 | Intron | XP_006717488.1 | |||
XM_011519416.2 | 23931 | Intron | XP_011517718.1 | |||
XM_017015928.1 | 23931 | Intron | XP_016871417.1 | |||
XM_017015929.1 | 23931 | UTR 3 | XP_016871418.1 | |||
XM_017015930.1 | 23931 | Intron | XP_016871419.1 | |||
XM_017015931.1 | 23931 | Intron | XP_016871420.1 | |||
XM_017015932.1 | 23931 | Intron | XP_016871421.1 | |||
XM_017015933.1 | 23931 | Intron | XP_016871422.1 | |||
XM_017015934.1 | 23931 | Intron | XP_016871423.1 |
LINC00202-1 - long intergenic non-protein coding RNA 202-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |