Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609589 MIM: 604584 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTUS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTUS1 - microtubule associated tumor suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001924.2 | 3013 | UTR 3 | NP_001001924.1 | |||
NM_001001925.2 | 3013 | UTR 3 | NP_001001925.1 | |||
NM_001001931.2 | 3013 | UTR 3 | NP_001001931.1 | |||
NM_001166393.1 | 3013 | UTR 3 | NP_001159865.1 | |||
NM_020749.4 | 3013 | UTR 3 | NP_065800.1 | |||
XM_005273577.3 | 3013 | UTR 3 | XP_005273634.1 | |||
XM_005273578.3 | 3013 | UTR 3 | XP_005273635.1 | |||
XM_005273579.3 | 3013 | UTR 3 | XP_005273636.1 | |||
XM_005273580.3 | 3013 | UTR 3 | XP_005273637.1 | |||
XM_005273581.3 | 3013 | UTR 3 | XP_005273638.1 | |||
XM_005273582.3 | 3013 | UTR 3 | XP_005273639.1 | |||
XM_005273583.4 | 3013 | Intron | XP_005273640.1 | |||
XM_011544595.2 | 3013 | UTR 3 | XP_011542897.1 | |||
XM_017013698.1 | 3013 | UTR 3 | XP_016869187.1 | |||
XM_017013699.1 | 3013 | UTR 3 | XP_016869188.1 | |||
XM_017013700.1 | 3013 | UTR 3 | XP_016869189.1 | |||
XM_017013701.1 | 3013 | UTR 3 | XP_016869190.1 | |||
XM_017013702.1 | 3013 | UTR 3 | XP_016869191.1 | |||
XM_017013703.1 | 3013 | Intron | XP_016869192.1 | |||
XM_017013704.1 | 3013 | UTR 3 | XP_016869193.1 |
PDGFRL - platelet derived growth factor receptor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006207.2 | 3013 | Intron | NP_006198.1 |
Set Membership: |
HapMap |