Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601643 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP2R5A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PPP2R5A - protein phosphatase 2 regulatory subunit B'alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | Intron | NP_001185791.1 | ||||
NM_018252.2 | Intron | NP_060722.2 | ||||
XM_005273177.3 | Intron | XP_005273234.1 | ||||
XM_011509717.2 | Intron | XP_011508019.1 | ||||
XM_011509718.2 | Intron | XP_011508020.1 | ||||
XM_017001634.1 | Intron | XP_016857123.1 | ||||
XM_017001635.1 | Intron | XP_016857124.1 | ||||
XM_017001636.1 | Intron | XP_016857125.1 | ||||
XM_017001637.1 | Intron | XP_016857126.1 |