Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTAAATACCCCAGGGTTTAAACT[A/G]ATTTATTCTTCTGTTACTTAGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608172 MIM: 163910 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHDDS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DHDDS - dehydrodolichyl diphosphate synthase subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMGN2 - high mobility group nucleosomal binding domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005517.3 | Intron | NP_005508.1 |
LOC101928324 - uncharacterized LOC101928324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |