Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGCAGACACTTCCTAACAAAGAA[C/T]GCAATAAGTCACTTTCGAAAGAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609780 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPVL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CPVL - carboxypeptidase, vitellogenic like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019029.2 | Intron | NP_061902.2 | ||||
NM_031311.3 | Intron | NP_112601.3 | ||||
XM_011515437.1 | Intron | XP_011513739.1 | ||||
XM_017012366.1 | Intron | XP_016867855.1 |
LOC100506497 - uncharacterized LOC100506497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |