Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602842 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMNN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GMNN - geminin, DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251989.1 | 23 | UTR 5 | NP_001238918.1 | |||
NM_001251990.1 | 23 | UTR 5 | NP_001238919.1 | |||
NM_001251991.1 | 23 | Intron | NP_001238920.1 | |||
NM_015895.4 | 23 | UTR 5 | NP_056979.1 | |||
XM_005249159.1 | 23 | Intron | XP_005249216.1 | |||
XM_011514651.2 | 23 | Intron | XP_011512953.1 |