Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612205 MIM: 163729 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG9B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATG9B - autophagy related 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317056.1 | 3404 | Intron | NP_001303985.1 | |||
XM_011516065.1 | 3404 | Intron | XP_011514367.1 | |||
XM_011516066.2 | 3404 | Intron | XP_011514368.1 |
NOS3 - nitric oxide synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000603.4 | 3404 | Silent Mutation | GTC,GTT | V,V 1117 | NP_000594.2 | |
NM_001160109.1 | 3404 | Intron | NP_001153581.1 | |||
NM_001160110.1 | 3404 | Intron | NP_001153582.1 | |||
NM_001160111.1 | 3404 | Intron | NP_001153583.1 | |||
XM_006716002.3 | 3404 | Intron | XP_006716065.1 | |||
XM_017012232.1 | 3404 | Silent Mutation | GTC,GTT | V,V 1117 | XP_016867721.1 | |
XM_017012233.1 | 3404 | Silent Mutation | GTC,GTT | V,V 911 | XP_016867722.1 | |
XM_017012234.1 | 3404 | Intron | XP_016867723.1 |
Set Membership: |
HapMap |