Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGCTAGTATAAAAGAAAGCATCA[A/T]TTAGAAAAAAAGCTTTTTACCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614797 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PELI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PELI1 - pellino E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020651.3 | Intron | NP_065702.2 | ||||
XM_005264437.3 | Intron | XP_005264494.1 | ||||
XM_011532994.2 | Intron | XP_011531296.1 | ||||
XM_011532996.2 | Intron | XP_011531298.1 | ||||
XM_011532997.2 | Intron | XP_011531299.1 | ||||
XM_017004520.1 | Intron | XP_016860009.1 | ||||
XM_017004521.1 | Intron | XP_016860010.1 |