Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603871 MIM: 123834 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BYSL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BYSL - bystin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136017.3 | Intron | NP_001129489.1 | ||||
NM_001136125.2 | Intron | NP_001129597.1 | ||||
NM_001136126.2 | Intron | NP_001129598.1 | ||||
NM_001287427.1 | Intron | NP_001274356.1 | ||||
NM_001287434.1 | Intron | NP_001274363.1 | ||||
NM_001760.4 | Intron | NP_001751.1 | ||||
XM_011514971.1 | Intron | XP_011513273.1 | ||||
XM_011514972.1 | Intron | XP_011513274.1 |
Set Membership: |
Validated |