Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCTATTATTTGAAGTACAGAA[C/T]TTAGAATATATTCTTAAAAATACAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615383 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FIGNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FIGNL1 - fidgetin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042762.2 | 2508 | UTR 3 | NP_001036227.1 | |||
NM_001287492.1 | 2508 | UTR 3 | NP_001274421.1 | |||
NM_001287493.1 | 2508 | UTR 3 | NP_001274422.1 | |||
NM_001287494.1 | 2508 | UTR 3 | NP_001274423.1 | |||
NM_001287495.1 | 2508 | UTR 3 | NP_001274424.1 | |||
NM_001287496.1 | 2508 | UTR 3 | NP_001274425.1 | |||
NM_022116.4 | 2508 | UTR 3 | NP_071399.2 | |||
XM_011515470.2 | 2508 | UTR 3 | XP_011513772.1 | |||
XM_017012501.1 | 2508 | UTR 3 | XP_016867990.1 | |||
XM_017012502.1 | 2508 | UTR 3 | XP_016867991.1 |
Set Membership: |
Validated |