Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCCGGTTCCCGGGGCGTCAGGT[C/T]GGCTGACAGTTCGGGGTCCCTGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAV2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAV2 - caveolin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206747.1 | 178 | Intron | NP_001193676.1 | |||
NM_001206748.1 | 178 | UTR 5 | NP_001193677.1 | |||
NM_001233.4 | 178 | Intron | NP_001224.1 | |||
NM_198212.2 | 178 | Intron | NP_937855.1 |