Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGGCACTCGAGCTCCACGAGGA[C/T]GGAGGTGACGCGCTGGGGCTTGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608832 | ||||||||||||||||||||
Literature Links: |
GREM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GREM2 - gremlin 2, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022469.3 | 671 | Missense Mutation | NP_071914.3 | |||
XM_005273226.3 | 671 | Missense Mutation | XP_005273283.1 | |||
XM_011544249.2 | 671 | Missense Mutation | XP_011542551.1 |