Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGATGGCAAGGATAGTACCAAT[C/G]GGAATAGACTTCTGAGCATCTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604878 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC12A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC12A6 - solute carrier family 12 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042494.1 | 1670 | Silent Mutation | CCC,CCG | P,P 458 | NP_001035959.1 | |
NM_001042495.1 | 1670 | Silent Mutation | CCC,CCG | P,P 458 | NP_001035960.1 | |
NM_001042496.1 | 1670 | Silent Mutation | CCC,CCG | P,P 508 | NP_001035961.1 | |
NM_001042497.1 | 1670 | Silent Mutation | CCC,CCG | P,P 502 | NP_001035962.1 | |
NM_005135.2 | 1670 | Silent Mutation | CCC,CCG | P,P 466 | NP_005126.1 | |
NM_133647.1 | 1670 | Silent Mutation | CCC,CCG | P,P 517 | NP_598408.1 | |
XM_006720793.3 | 1670 | Silent Mutation | CCC,CCG | P,P 468 | XP_006720856.1 | |
XM_011522269.2 | 1670 | Silent Mutation | CCC,CCG | P,P 517 | XP_011520571.1 |
Set Membership: |
HapMap |