Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGACACCAGCTGCACAGTGTCTG[T/C]GTGCTCAGCCAGGTGCTGGAAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAM8 - ADAM metallopeptidase domain 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBGCP2 - tubulin gamma complex associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256617.1 | 2798 | Missense Mutation | NP_001243546.1 | |||
NM_001256618.1 | 2798 | Missense Mutation | NP_001243547.1 | |||
NM_006659.3 | 2798 | Missense Mutation | NP_006650.1 | |||
XM_017015595.1 | 2798 | Missense Mutation | XP_016871084.1 | |||
XM_017015596.1 | 2798 | Missense Mutation | XP_016871085.1 |
Set Membership: |
HapMap |