Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGACAAACTTGTTCACATACA[A/G]AAATGGGATGGCAAAGAAACAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602965 MIM: 610728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FABP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FABP7 - fatty acid binding protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319039.1 | 349 | Silent Mutation | CAA,CAG | Q,Q 96 | NP_001305968.1 | |
NM_001319041.1 | 349 | UTR 3 | NP_001305970.1 | |||
NM_001319042.1 | 349 | Silent Mutation | CAA,CAG | Q,Q 92 | NP_001305971.1 | |
NM_001446.4 | 349 | Silent Mutation | CAA,CAG | Q,Q 96 | NP_001437.1 |
SMPDL3A - sphingomyelin phosphodiesterase acid like 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |