Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGTTCTCGGCTGGCAGGACGA[C/T]GCACGTCACCCCTGCGCGCTTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605490 | ||||||||||||||||||||
Literature Links: |
HSD11B1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSD11B1L - hydroxysteroid 11-beta dehydrogenase 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LONP1 - lon peptidase 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276479.1 | 2569 | Missense Mutation | ATC,GTC | I,V 846 | NP_001263408.1 | |
NM_001276480.1 | 2569 | Missense Mutation | ATC,GTC | I,V 714 | NP_001263409.1 | |
NM_004793.3 | 2569 | Missense Mutation | ATC,GTC | I,V 910 | NP_004784.2 | |
XM_011528441.2 | 2569 | Intron | XP_011526743.1 |
RPL36 - ribosomal protein L36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015414.3 | 2569 | Intron | NP_056229.2 | |||
NM_033643.2 | 2569 | Intron | NP_378669.1 |