Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCAGAGGCAGGTGCTGCAAGGT[A/G]GGGCCGAGGCGGGCAGGTGCCCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPAG6 - sperm associated antigen 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253854.1 | 195 | Intron | NP_001240783.1 | |||
NM_001253855.1 | 195 | UTR 5 | NP_001240784.1 | |||
NM_012443.3 | 195 | Intron | NP_036575.1 | |||
NM_172242.2 | 195 | Intron | NP_758442.1 | |||
XM_005252645.1 | 195 | Intron | XP_005252702.1 | |||
XM_005252646.1 | 195 | Intron | XP_005252703.1 |