Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAGCGGGTACGAGTCGGTGCT[C/G]TGTGTCAAGCCTGACGTCCACGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611624 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NECAP2 - NECAP endocytosis associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145277.1 | 120 | Silent Mutation | CTC,CTG | L,L 10 | NP_001138749.1 | |
NM_001145278.1 | 120 | Intron | NP_001138750.1 | |||
NM_018090.4 | 120 | Silent Mutation | CTC,CTG | L,L 10 | NP_060560.1 | |
XM_011541767.2 | 120 | UTR 5 | XP_011540069.1 | |||
XM_017001721.1 | 120 | Silent Mutation | CTC,CTG | L,L 10 | XP_016857210.1 |
SPATA21 - spermatogenesis associated 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |