Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATTTGCAGGCAAACATCCCACA[A/G]TCACTTCCATTCATCTGCTGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612157 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SENP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SENP1 - SUMO1/sentrin specific peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267594.1 | 2382 | Silent Mutation | GAC,GAT | D,D 602 | NP_001254523.1 | |
NM_001267595.1 | 2382 | Silent Mutation | GAC,GAT | D,D 602 | NP_001254524.1 | |
XM_006719361.3 | 2382 | Silent Mutation | GAC,GAT | D,D 634 | XP_006719424.1 | |
XM_006719362.2 | 2382 | Silent Mutation | GAC,GAT | D,D 403 | XP_006719425.1 | |
XM_011538244.2 | 2382 | Silent Mutation | GAC,GAT | D,D 621 | XP_011536546.1 | |
XM_011538245.2 | 2382 | Silent Mutation | GAC,GAT | D,D 618 | XP_011536547.1 | |
XM_017019229.1 | 2382 | Silent Mutation | GAC,GAT | D,D 665 | XP_016874718.1 | |
XM_017019230.1 | 2382 | Silent Mutation | GAC,GAT | D,D 652 | XP_016874719.1 | |
XM_017019231.1 | 2382 | Silent Mutation | GAC,GAT | D,D 633 | XP_016874720.1 | |
XM_017019232.1 | 2382 | Silent Mutation | GAC,GAT | D,D 633 | XP_016874721.1 | |
XM_017019233.1 | 2382 | Silent Mutation | GAC,GAT | D,D 595 | XP_016874722.1 | |
XM_017019234.1 | 2382 | Silent Mutation | GAC,GAT | D,D 595 | XP_016874723.1 | |
XM_017019235.1 | 2382 | Silent Mutation | GAC,GAT | D,D 595 | XP_016874724.1 | |
XM_017019236.1 | 2382 | Silent Mutation | GAC,GAT | D,D 589 | XP_016874725.1 | |
XM_017019237.1 | 2382 | Silent Mutation | GAC,GAT | D,D 589 | XP_016874726.1 | |
XM_017019238.1 | 2382 | Silent Mutation | GAC,GAT | D,D 403 | XP_016874727.1 | |
XM_017019239.1 | 2382 | Silent Mutation | GAC,GAT | D,D 403 | XP_016874728.1 | |
XM_017019240.1 | 2382 | Silent Mutation | GAC,GAT | D,D 390 | XP_016874729.1 |