Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGGGACAGCAGAAGTGAGACTG[A/G]CAGTATGTCTTAGCTCAGGATGTGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611433 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGOHB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAGOHB - mago homolog B, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYK1 - serine/threonine/tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018423.2 | 1986 | UTR 3 | NP_060893.2 | |||
XM_005253417.2 | 1986 | UTR 3 | XP_005253474.1 | |||
XM_011520736.1 | 1986 | UTR 3 | XP_011519038.1 | |||
XM_011520737.2 | 1986 | UTR 3 | XP_011519039.1 | |||
XM_011520738.2 | 1986 | UTR 3 | XP_011519040.1 | |||
XM_017019579.1 | 1986 | UTR 3 | XP_016875068.1 |
Set Membership: |
HapMap |