Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611987 MIM: 609511 | |||||||||||||||||||||||
Literature Links: |
MRPS25 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MRPS25 - mitochondrial ribosomal protein S25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBSN - rabenosyn, RAB effector | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302378.1 | 2625 | Missense Mutation | ATA,ATG | I,M 722 | NP_001289307.1 | |
NM_022340.3 | 2625 | Missense Mutation | ATA,ATG | I,M 722 | NP_071735.2 | |
XM_005265384.4 | 2625 | Missense Mutation | ATA,ATG | I,M 722 | XP_005265441.1 | |
XM_005265385.4 | 2625 | Missense Mutation | ATA,ATG | I,M 722 | XP_005265442.1 | |
XM_011534001.2 | 2625 | Missense Mutation | ATA,ATG | I,M 634 | XP_011532303.1 | |
XM_017007023.1 | 2625 | Missense Mutation | ATA,ATG | I,M 722 | XP_016862512.1 | |
XM_017007024.1 | 2625 | Missense Mutation | ATA,ATG | I,M 633 | XP_016862513.1 | |
XM_017007025.1 | 2625 | Missense Mutation | ATA,ATG | I,M 633 | XP_016862514.1 | |
XM_017007026.1 | 2625 | Missense Mutation | ATA,ATG | I,M 537 | XP_016862515.1 | |
XM_017007027.1 | 2625 | Missense Mutation | ATA,ATG | I,M 483 | XP_016862516.1 | |
XM_017007028.1 | 2625 | Missense Mutation | ATA,ATG | I,M 483 | XP_016862517.1 |