Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAACGGGACCCTGGTTCTGCCG[A/G]AAATGTGAATCTCAGGAGCGAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600328 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371763 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371763 - uncharacterized LOC105371763 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4734 - microRNA 4734 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MLLT6 - MLLT6, PHD finger domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005937.3 | 169 | Silent Mutation | CGA,CGG | R,R 52 | NP_005928.2 | |
XM_006721911.2 | 169 | Silent Mutation | CGA,CGG | R,R 52 | XP_006721974.1 | |
XM_017024648.1 | 169 | Intron | XP_016880137.1 | |||
XM_017024649.1 | 169 | Silent Mutation | CGA,CGG | R,R 52 | XP_016880138.1 | |
XM_017024650.1 | 169 | Silent Mutation | CGA,CGG | R,R 52 | XP_016880139.1 |