Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCCGCTGGAGCCCGGTGAGGGC[C/T]GGGGCCCCTCGATTCCCCCTGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609077 MIM: 602438 MIM: 605235 MIM: 603500 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXL8 - F-box and leucine rich repeat protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018378.2 | Intron | NP_060848.2 |
HSF4 - heat shock transcription factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040667.2 | Intron | NP_001035757.1 | ||||
NM_001538.3 | Intron | NP_001529.2 |
NOL3 - nucleolar protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRADD - TNFRSF1A associated via death domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |