Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613899 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FANCC - Fanconi anemia complementation group C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000136.2 | 2182 | UTR 3 | NP_000127.2 | |||
NM_001243743.1 | 2182 | UTR 3 | NP_001230672.1 | |||
NM_001243744.1 | 2182 | Intron | NP_001230673.1 | |||
XM_005251802.3 | 2182 | UTR 3 | XP_005251859.1 | |||
XM_006717001.2 | 2182 | UTR 3 | XP_006717064.1 | |||
XM_006717002.3 | 2182 | Intron | XP_006717065.1 | |||
XM_006717004.3 | 2182 | Intron | XP_006717067.1 | |||
XM_011518365.2 | 2182 | UTR 3 | XP_011516667.1 | |||
XM_011518366.2 | 2182 | Intron | XP_011516668.1 | |||
XM_011518367.2 | 2182 | UTR 3 | XP_011516669.1 | |||
XM_017014452.1 | 2182 | UTR 3 | XP_016869941.1 | |||
XM_017014453.1 | 2182 | UTR 3 | XP_016869942.1 | |||
XM_017014454.1 | 2182 | UTR 3 | XP_016869943.1 |
Set Membership: |
HapMap |