Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAACATAGGTAGTGGATAAAGGT[G/T]CTAGCATATGACACACTCCATTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600682 | ||||||||||||||||||||||||||||||||
Literature Links: |
AKR7A2P1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
AKR7A2P1 - aldo-keto reductase family 7 member A2 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A1 - solute carrier family 16 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166496.1 | 1403 | Missense Mutation | GAA,GCA | E,A 347 | NP_001159968.1 | |
NM_003051.3 | 1403 | Missense Mutation | GAA,GCA | E,A 347 | NP_003042.3 | |
XM_011542026.2 | 1403 | Intron | XP_011540328.1 | |||
XM_011542027.2 | 1403 | Intron | XP_011540329.1 |
Set Membership: |
DME Validated Inventoried |