Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGTATATTTGATTTCTTCAGCAG[C/T]GTTTAGAATTCCAGATAATTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137167 MIM: 601468 MIM: 616350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGCX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGCX - gamma-glutamyl carboxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAT2A - methionine adenosyltransferase 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005911.5 | Intron | NP_005902.1 |
PARTICL - promoter of MAT2A antisense radiation-induced circulating long non-coding RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |