Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTTAATTCTCATTTTCTTTCC[C/T]CTTTCACAGTGTGTAACCACTCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604333 MIM: 601664 MIM: 613403 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CIAO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CIAO1 - cytosolic iron-sulfur assembly component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004804.2 | Intron | NP_004795.1 |
SNRNP200 - small nuclear ribonucleoprotein U5 subunit 200 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM127 - transmembrane protein 127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |