Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCACTGGAGCTGGCCACTGTCC[G/T]TGGGCAGGATACACTGTGTTGGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604808 MIM: 611567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLRT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLRT3 - fibronectin leucine rich transmembrane protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013281.3 | 2299 | Missense Mutation | CAA,CAC | Q,H 378 | NP_037413.1 | |
NM_198391.2 | 2299 | Missense Mutation | CAA,CAC | Q,H 378 | NP_938205.1 | |
XM_005260682.4 | 2299 | Missense Mutation | CAA,CAC | Q,H 378 | XP_005260739.1 | |
XM_011529204.2 | 2299 | Missense Mutation | CAA,CAC | Q,H 378 | XP_011527506.1 | |
XM_011529205.2 | 2299 | Missense Mutation | CAA,CAC | Q,H 378 | XP_011527507.1 |
MACROD2 - MACRO domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033087.1 | 2299 | Intron | NP_001028259.1 | |||
NM_080676.5 | 2299 | Intron | NP_542407.2 | |||
XM_017027675.1 | 2299 | Intron | XP_016883164.1 | |||
XM_017027676.1 | 2299 | Intron | XP_016883165.1 | |||
XM_017027677.1 | 2299 | Intron | XP_016883166.1 |