Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGGATAGGGCCTCTGCCAGAGA[A/G]GAGGCTCCCTCCCCACCTCAGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ILVBL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ILVBL - ilvB acetolactate synthase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006844.4 | Intron | NP_006835.2 | ||||
XM_005259717.4 | Intron | XP_005259774.1 | ||||
XM_011527651.2 | Intron | XP_011525953.1 |
SYDE1 - synapse defective Rho GTPase homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300910.1 | Intron | NP_001287839.1 | ||||
NM_033025.5 | Intron | NP_149014.3 |