Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCATCCGTGGGGGGGCGCCGGCT[C/T]GGGCGTGCGGGGAGTGTCCGCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601047 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CAV1 - caveolin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172895.1 | 112 | UTR 5 | NP_001166366.1 | |||
NM_001172896.1 | 112 | Intron | NP_001166367.1 | |||
NM_001172897.1 | 112 | Intron | NP_001166368.1 | |||
NM_001753.4 | 112 | Intron | NP_001744.2 |
Set Membership: |
HapMap |