Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTCCCTGATGCTAATTTTGT[A/G]AGAAGGTAAAAGGACACCCTGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607568 MIM: 251170 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMAB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MMAB - methylmalonic aciduria (cobalamin deficiency) cblB type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVK - mevalonate kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000431.3 | Intron | NP_000422.1 | ||||
NM_001114185.2 | Intron | NP_001107657.1 | ||||
NM_001301182.1 | Intron | NP_001288111.1 | ||||
XM_011538372.2 | Intron | XP_011536674.2 | ||||
XM_017019313.1 | Intron | XP_016874802.1 | ||||
XM_017019314.1 | Intron | XP_016874803.1 |
Set Membership: |
Validated |