Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGGACAGCTATGAGGAGCAGC[A/G]CAGTAAGGGATGATGCCGGGCCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603889 MIM: 605466 | ||||||||||||||||||||
Literature Links: |
SIRPB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SIRPB1 - signal regulatory protein beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRPG - signal regulatory protein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039508.1 | 835 | Missense Mutation | GCG,GTG | A,V 257 | NP_001034597.1 | |
NM_018556.3 | 835 | Missense Mutation | GCG,GTG | A,V 368 | NP_061026.2 | |
NM_080816.2 | 835 | Missense Mutation | GCG,GTG | A,V 151 | NP_543006.2 | |
XM_005260749.3 | 835 | Intron | XP_005260806.1 | |||
XM_011529286.2 | 835 | Missense Mutation | GCG,GTG | A,V 335 | XP_011527588.1 | |
XM_011529287.2 | 835 | Intron | XP_011527589.1 |
SIRPG-AS1 - SIRPG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |