Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATTGGGATCTGAGGGGTCCAAA[C/T]TGAACAAGTGGGAAGTGTACATTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESF1 - ESF1 nucleolar pre-rRNA processing protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276380.1 | 3875 | Missense Mutation | AAT,AGT | N,S 772 | NP_001263309.1 | |
NM_016649.3 | 3875 | Missense Mutation | AAT,AGT | N,S 772 | NP_057733.2 | |
XM_017027874.1 | 3875 | Missense Mutation | AAT,AGT | N,S 772 | XP_016883363.1 |