Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCTGACATAGAAAAGGTTAAC[G/C]AGAATCAGGTAAGGAGAAAGTTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300275 | ||||||||||||||||||||
Literature Links: |
NSDHL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NSDHL - NAD(P) dependent steroid dehydrogenase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001129765.1 | 226 | Missense Mutation | CAG,GAG | Q,E 34 | NP_001123237.1 | |
NM_015922.2 | 226 | Missense Mutation | CAG,GAG | Q,E 34 | NP_057006.1 | |
XM_011531178.2 | 226 | Missense Mutation | CAG,GAG | Q,E 34 | XP_011529480.1 | |
XM_017029564.1 | 226 | Missense Mutation | CAG,GAG | Q,E 50 | XP_016885053.1 |