Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCAAGAGGGCGATGAGGGCGC[G/C]GGGGCTGAGGGAGGCGGCCATGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609920 | ||||||||||||||||||||
Literature Links: |
CDH22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH22 - cadherin 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021248.2 | 2475 | Missense Mutation | CGC,GGC | R,G 625 | NP_067071.1 | |
XM_011528994.2 | 2475 | Missense Mutation | CGC,GGC | R,G 625 | XP_011527296.1 | |
XM_011528995.1 | 2475 | Missense Mutation | CGC,GGC | R,G 504 | XP_011527297.1 |