Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600214 MIM: 603099 MIM: 176311 MIM: 602677 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGER PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AGER - advanced glycosylation end product-specific receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136.4 | Intron | NP_001127.1 | ||||
NM_001206929.1 | Intron | NP_001193858.1 | ||||
NM_001206932.1 | Intron | NP_001193861.1 | ||||
NM_001206934.1 | Intron | NP_001193863.1 | ||||
NM_001206936.1 | Intron | NP_001193865.1 | ||||
NM_001206940.1 | Intron | NP_001193869.1 | ||||
NM_001206954.1 | Intron | NP_001193883.1 | ||||
NM_001206966.1 | Intron | NP_001193895.1 | ||||
NM_172197.2 | Intron | NP_751947.1 | ||||
XM_017010328.1 | Intron | XP_016865817.1 |
AGPAT1 - 1-acylglycerol-3-phosphate O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPSM3 - G-protein signaling modulator 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6833 - microRNA 6833 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PBX2 - PBX homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF5 - ring finger protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |