Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600266 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC11A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC11A1 - solute carrier family 11 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000578.3 | 718 | Silent Mutation | CTG,TTG | L,L 39 | NP_000569.3 | |
XM_005246793.3 | 718 | Intron | XP_005246850.1 | |||
XM_005246794.3 | 718 | UTR 5 | XP_005246851.1 | |||
XM_006712709.3 | 718 | Intron | XP_006712772.1 | |||
XM_006712710.3 | 718 | Intron | XP_006712773.1 | |||
XM_006712711.3 | 718 | Intron | XP_006712774.1 | |||
XM_011511684.2 | 718 | UTR 5 | XP_011509986.1 | |||
XM_011511685.2 | 718 | Intron | XP_011509987.1 | |||
XM_017004765.1 | 718 | Silent Mutation | CTG,TTG | L,L 39 | XP_016860254.1 | |
XM_017004766.1 | 718 | UTR 5 | XP_016860255.1 | |||
XM_017004767.1 | 718 | Silent Mutation | CTG,TTG | L,L 39 | XP_016860256.1 |
Set Membership: |
HapMap |