Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCGGCCTCGGTCCCCCGGGACT[A/G]GACTGCCCCTTCCGCCTGGCGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610660 MIM: 609771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLYR1 - glyoxylate reductase 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308096.1 | 397 | Intron | NP_001295025.1 | |||
NM_001324096.1 | 397 | Intron | NP_001311025.1 | |||
NM_001324097.1 | 397 | Intron | NP_001311026.1 | |||
NM_001324098.1 | 397 | Intron | NP_001311027.1 | |||
NM_032569.3 | 397 | Intron | NP_115958.2 | |||
XM_005255639.4 | 397 | Intron | XP_005255696.2 | |||
XM_011522716.2 | 397 | Intron | XP_011521018.1 | |||
XM_011522717.1 | 397 | Intron | XP_011521019.1 | |||
XM_017023786.1 | 397 | Intron | XP_016879275.1 | |||
XM_017023787.1 | 397 | Intron | XP_016879276.1 | |||
XM_017023788.1 | 397 | Intron | XP_016879277.1 | |||
XM_017023789.1 | 397 | Intron | XP_016879278.1 |
UBN1 - ubinuclein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079514.2 | 397 | Intron | NP_001072982.1 | |||
NM_001288656.1 | 397 | Intron | NP_001275585.1 | |||
XM_005255277.3 | 397 | Intron | XP_005255334.1 | |||
XM_005255278.3 | 397 | UTR 5 | XP_005255335.1 | |||
XM_005255279.3 | 397 | Intron | XP_005255336.1 | |||
XM_011522465.1 | 397 | Intron | XP_011520767.1 | |||
XM_011522466.2 | 397 | Intron | XP_011520768.1 | |||
XM_011522467.1 | 397 | Intron | XP_011520769.1 | |||
XM_017023189.1 | 397 | Intron | XP_016878678.1 | |||
XM_017023190.1 | 397 | Intron | XP_016878679.1 | |||
XM_017023191.1 | 397 | Intron | XP_016878680.1 | |||
XM_017023192.1 | 397 | Intron | XP_016878681.1 |