Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCACCAGGGGGCTGGTCCTGCCT[C/G]ACTCGAGATGACAGCTCAGCTTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS3 - ankyrin repeat and sterile alpha motif domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242929.1 | 1718 | Silent Mutation | GTC,GTG | V,V 478 | NP_001229858.1 | |
NM_001308089.1 | 1718 | Silent Mutation | GTC,GTG | V,V 456 | NP_001295018.1 | |
NM_001324129.1 | 1718 | Silent Mutation | GTC,GTG | V,V 412 | NP_001311058.1 | |
NM_001324130.1 | 1718 | Silent Mutation | GTC,GTG | V,V 295 | NP_001311059.1 | |
NM_133450.3 | 1718 | Silent Mutation | GTC,GTG | V,V 585 | NP_597707.1 | |
XM_005255119.3 | 1718 | Silent Mutation | GTC,GTG | V,V 456 | XP_005255176.1 | |
XM_005255121.3 | 1718 | Silent Mutation | GTC,GTG | V,V 379 | XP_005255178.1 | |
XM_011522372.1 | 1718 | Silent Mutation | GTC,GTG | V,V 597 | XP_011520674.1 | |
XM_011522373.1 | 1718 | Silent Mutation | GTC,GTG | V,V 585 | XP_011520675.1 | |
XM_011522374.1 | 1718 | Silent Mutation | GTC,GTG | V,V 478 | XP_011520676.1 | |
XM_011522378.1 | 1718 | Silent Mutation | GTC,GTG | V,V 379 | XP_011520680.1 | |
XM_017022935.1 | 1718 | Silent Mutation | GTC,GTG | V,V 490 | XP_016878424.1 | |
XM_017022936.1 | 1718 | Silent Mutation | GTC,GTG | V,V 379 | XP_016878425.1 |
MGRN1 - mahogunin ring finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT16L1 - nudix hydrolase 16 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |