Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTATTTGTTTCTTTTAGTGCATC[A/G]CCAAAATATAAATCAACATTAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF19A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF19A - ring finger protein 19A, RBR E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280539.1 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | NP_001267468.1 | |
NM_015435.4 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | NP_056250.3 | |
NM_183419.3 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | NP_904355.1 | |
XM_005250853.3 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | XP_005250910.1 | |
XM_011516956.2 | 2612 | Silent Mutation | GGC,GGT | G,G 759 | XP_011515258.1 | |
XM_017013302.1 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | XP_016868791.1 | |
XM_017013303.1 | 2612 | Silent Mutation | GGC,GGT | G,G 813 | XP_016868792.1 | |
XM_017013304.1 | 2612 | Silent Mutation | GGC,GGT | G,G 759 | XP_016868793.1 | |
XM_017013305.1 | 2612 | Silent Mutation | GGC,GGT | G,G 503 | XP_016868794.1 |