Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGCGACTGCACGGTGCTTGTAG[G/C]GGGGGCCGGCCTCTGGGAGGGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603464 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU)
|
||||||
EAS - Not Available | African American
|
YRI (Yoruba)
|
||||||
SAS - Not Available | Japanese
|
JPT (Japanese)
|
||||||
AFR - Not Available | Chinese
|
CHB (Han Chinese)
|
||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CDK10 - cyclin dependent kinase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098533.2 | Intron | NP_001092003.2 | ||||
NM_001160367.1 | Intron | NP_001153839.1 | ||||
NM_052987.3 | Intron | NP_443713.2 | ||||
NM_052988.4 | Intron | NP_443714.3 | ||||
XM_006721308.2 | Intron | XP_006721371.1 | ||||
XM_006721310.2 | Intron | XP_006721373.1 | ||||
XM_011523405.2 | Intron | XP_011521707.1 | ||||
XM_011523406.2 | Intron | XP_011521708.1 | ||||
XM_011523407.2 | Intron | XP_011521709.1 | ||||
XM_011523408.2 | Intron | XP_011521710.1 | ||||
XM_011523410.2 | Intron | XP_011521712.1 | ||||
XM_011523411.2 | Intron | XP_011521713.1 | ||||
XM_011523412.1 | Intron | XP_011521714.1 | ||||
XM_011523413.2 | Intron | XP_011521715.1 | ||||
XM_011523414.2 | Intron | XP_011521716.1 | ||||
XM_011523415.2 | Intron | XP_011521717.1 | ||||
XM_011523416.2 | Intron | XP_011521718.1 | ||||
XM_011523417.2 | Intron | XP_011521719.1 | ||||
XM_017023806.1 | Intron | XP_016879295.1 | ||||
XM_017023807.1 | Intron | XP_016879296.1 | ||||
XM_017023808.1 | Intron | XP_016879297.1 | ||||
XM_017023809.1 | Intron | XP_016879298.1 | ||||
XM_017023810.1 | Intron | XP_016879299.1 |
SPATA2L - spermatogenesis associated 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |