Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCACATTGATGTAATCCAGGAT[C/T]GTGCTGTGCCGGGAGAACCTGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606091 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100129083 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100129083 - uncharacterized LOC100129083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIGLEC10 - sialic acid binding Ig like lectin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171156.1 | 1542 | Silent Mutation | ACA,ACG | T,T 535 | NP_001164627.1 | |
NM_001171157.1 | 1542 | Intron | NP_001164628.1 | |||
NM_001171158.1 | 1542 | Silent Mutation | ACA,ACG | T,T 450 | NP_001164629.1 | |
NM_001171159.1 | 1542 | Silent Mutation | ACA,ACG | T,T 440 | NP_001164630.1 | |
NM_001171161.1 | 1542 | Silent Mutation | ACA,ACG | T,T 350 | NP_001164632.1 | |
NM_001322105.1 | 1542 | Silent Mutation | ACA,ACG | T,T 408 | NP_001309034.1 | |
NM_033130.4 | 1542 | Silent Mutation | ACA,ACG | T,T 593 | NP_149121.2 | |
XM_005259366.4 | 1542 | Silent Mutation | ACA,ACG | T,T 503 | XP_005259423.1 | |
XM_005259367.3 | 1542 | Silent Mutation | ACA,ACG | T,T 445 | XP_005259424.1 | |
XM_011527439.1 | 1542 | Silent Mutation | ACA,ACG | T,T 545 | XP_011525741.1 |