Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACCATGTCACTGGTCTACCTG[G/C]TCTGGGAGCCTGTCAAGAAAAGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611486 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYCE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYCE1 - synaptonemal complex central element protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143763.1 | 1033 | Missense Mutation | CCA,GCA | P,A 310 | NP_001137235.1 | |
NM_001143764.1 | 1033 | Intron | NP_001137236.1 | |||
NM_130784.2 | 1033 | Missense Mutation | CCA,GCA | P,A 274 | NP_570140.1 |