Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605222 | |||||||||||||||||||||||
Literature Links: |
KCNMB3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KCNMB3 - potassium calcium-activated channel subfamily M regulatory beta subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163677.1 | 669 | Missense Mutation | ACT,GCT | T,A 51 | NP_001157149.1 | |
NM_014407.3 | 669 | Missense Mutation | ACT,GCT | T,A 53 | NP_055222.3 | |
NM_171828.2 | 669 | Missense Mutation | ACT,GCT | T,A 51 | NP_741979.1 | |
NM_171829.2 | 669 | Missense Mutation | ACT,GCT | T,A 31 | NP_741980.1 | |
NM_171830.1 | 669 | Missense Mutation | ACT,GCT | T,A 49 | NP_741981.1 |