Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCGCCCCAGATGGCTTCTGCTGC[C/T]TGCTCCATGGACCCCATCGACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB3L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CREB3L3 - cAMP responsive element binding protein 3 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271995.1 | 189 | Silent Mutation | GCC,GCT | A,A 14 | NP_001258924.1 | |
NM_001271996.1 | 189 | Silent Mutation | GCC,GCT | A,A 14 | NP_001258925.1 | |
NM_001271997.1 | 189 | Silent Mutation | GCC,GCT | A,A 14 | NP_001258926.1 | |
NM_032607.2 | 189 | Silent Mutation | GCC,GCT | A,A 14 | NP_115996.1 |