Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGCTGCGCAGTGGTGGAGGGG[A/C]CGACGCCCTCGTGGTGCTGCTCAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf210 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf210 - chromosome 1 open reading frame 210 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM125 - transmembrane protein 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320244.1 | 1067 | Missense Mutation | GAC,GCC | D,A 112 | NP_001307173.1 | |
NM_144626.2 | 1067 | Missense Mutation | GAC,GCC | D,A 112 | NP_653227.1 | |
XM_005270467.3 | 1067 | Missense Mutation | GAC,GCC | D,A 112 | XP_005270524.1 | |
XM_011540705.2 | 1067 | Missense Mutation | GAC,GCC | D,A 112 | XP_011539007.1 |