Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCACCAGGACGGAGAATGTGACC[A/G]TTGGGGGCTACTACCCAGGGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105377805 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105377805 - basic salivary proline-rich protein 4-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM255B - transmembrane protein 255B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182614.2 | 190 | Missense Mutation | ATT,GTT | I,V 55 | NP_872420.1 | |
XM_005272413.3 | 190 | Missense Mutation | ATT,GTT | I,V 55 | XP_005272470.1 | |
XM_005272415.2 | 190 | UTR 5 | XP_005272472.1 | |||
XM_006719986.2 | 190 | Intron | XP_006720049.1 | |||
XM_017020558.1 | 190 | Missense Mutation | ATT,GTT | I,V 55 | XP_016876047.1 | |
XM_017020559.1 | 190 | UTR 5 | XP_016876048.1 | |||
XM_017020560.1 | 190 | Missense Mutation | ATT,GTT | I,V 55 | XP_016876049.1 |