Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTGGCTTCATTTTTCTTCGATC[A/G]TGGACGAAAAATATCAGCTTGAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605188 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR85 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPR85 - G protein-coupled receptor 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146265.1 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | NP_001139737.1 | |
NM_001146266.1 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | NP_001139738.1 | |
NM_001146267.1 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | NP_001139739.1 | |
NM_018970.6 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | NP_061843.3 | |
XM_005250451.2 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | XP_005250508.1 | |
XM_017012352.1 | 1319 | Silent Mutation | CAC,CAT | H,H 210 | XP_016867841.1 |
Set Membership: |
HapMap |